Learn More
Strem, An Ascensus Company CAS# 15635-87-7. 250mg. Iridium(III) acetylacetonate, 98%. MFCD00015353
SDP
Catalog No. 5090113131
Encompass
CAS# 15635-87-7. 250mg. Iridium(III) acetylacetonate, 98%. MFCD00015353. Molecular Weight: 489.53. Molecular Formula: Ir(CH3COCHCOCH3)3. Color/form: orange xtl. Strem# 77-1500. http://www.strem.com/catalog/v/77-1500/
Catalog No. 50-901-13131 Supplier Strem, An Ascensus Company Supplier No. 771500250MG
CAS# 15635-87-7. 250mg. Iridium(III) acetylacetonate, 98%. MFCD00015353. Molecular Weight: 489.53. Molecular Formula: Ir(CH3COCHCOCH3)3. Color/form: orange xtl. Strem# 77-1500. http://www.strem.com/catalog/v/77-1500/