Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Iridium(III) chloride

Catalog No. 502907992
Encompass
Change view
Click to view available options
Quantity:
2 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-290-7992 2 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-290-7992 Supplier Sigma Aldrich Supplier No. 3368072G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Iridium(III) chloride
TRUSTED_SUSTAINABILITY

Specifications

CAS 10025-83-9
Melting Point 763°C
Molecular Formula Cl3Ir
Linear Formula IrCl3
Quantity 2 g
MDL Number MFCD00011063
Synonym Iridium trichloride; Iridium(3+) chloride; Trichloroiridium
Molecular Weight (g/mol) 298.58
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.