Learn More
Iridium(III) chloride, anhydrous, Premion™, 99.99% (metals basis), Ir 63.9% min, Thermo Scientific Chemicals
Description
Iridium(III) chloride is used in iridium electroplating, in making the standard meter bar of paris and crucibles. It is used for high temperature electrical contacts, as hardening agent, in tipping pens and compass bearings and Pt/Ir spark plugs. It is used for medical, surgical or other patient oriented applications, MMO (mixed metal oxide) titanium electrode coatings and as a starting material for heterogeneous catalysts.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 763°C (decomposition) |
Color | Brown to Green |
Physical Form | Powder |
Quantity | 5 g |
Assay Percent Range | 99.99% (Metal basis) |
Solubility Information | Insoluble in water. |
Formula Weight | 298.56 |
Sensitivity | Hygroscopic |
Appearance | Green to brown powder |
Chemical Name or Material | Iridium(III) chloride |
Safety and Handling
Hazard Category | H290-H314-H335 |
Precautionary Statement | P234-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c |
EINECSNumber | 233-044-6 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.