Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iridium(III) chloride, anhydrous, Premion™, 99.99% (metals basis), Ir 63.9% min

Catalog No. AA4338003
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4338003 1 g
AA4338006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4338003 Supplier Thermo Scientific Chemicals Supplier No. 04338003
Add to Cart

CAS: 10025-83-9 | Cl3Ir | 298.567 g/mol

Iridium(III) chloride is used in iridium electroplating, in making the standard meter bar of paris and crucibles. It is used for high temperature electrical contacts, as hardening agent, in tipping pens and compass bearings and Pt/Ir spark plugs. It is used for medical, surgical or other patient oriented applications, MMO (mixed metal oxide) titanium electrode coatings and as a starting material for heterogeneous catalysts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10025-83-9
Molecular Formula Cl3Ir
Molecular Weight (g/mol) 298.567
MDL Number MFCD00011063
InChI Key DANYXEHCMQHDNX-UHFFFAOYSA-K
PubChem CID 25563
IUPAC Name trichloroiridium
SMILES Cl[Ir](Cl)Cl

Specifications

Melting Point 763°C (decomposition)
Color Brown to Green
Physical Form Powder
Quantity 1 g
Assay Percent Range 99.99% (Metal basis)
Solubility Information Insoluble in water.
Formula Weight 298.56
Sensitivity Hygroscopic
Appearance Green to brown powder
Chemical Name or Material Iridium(III) chloride
Hazard Category H290-H314-H335
Precautionary Statement P234-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c
EINECSNumber 233-044-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.