Learn More
Strem, An Ascensus Company CAS# 14996-61-3. 5g. Iridium(III) chloride hydrate (99.9%-Ir). MFCD00149749
SDP
Catalog No. 5090113148
Encompass
CAS# 14996-61-3. 5g. Iridium(III) chloride hydrate (99.9%-Ir). MFCD00149749. Molecular Weight: 298.58. Molecular Formula: IrCl3∙XH2O. Color/form: black xtl. Strem# 77-3000. http://www.strem.com/catalog/v/77-3000/
Catalog No. 50-901-13148 Supplier Strem, An Ascensus Company Supplier No. 7730005G
CAS# 14996-61-3. 5g. Iridium(III) chloride hydrate (99.9%-Ir). MFCD00149749. Molecular Weight: 298.58. Molecular Formula: IrCl3∙XH2O. Color/form: black xtl. Strem# 77-3000. http://www.strem.com/catalog/v/77-3000/