Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Iridium(III) chloride hydrate | 14996-61-3 | MFCD00149749 | 98.0% | 298.58 g/mol (anhydrous basis) | IrCl3 · xH2O | 1 G
SDP

Catalog No. 5000300115
Encompass_Preferred

Iridium(III) chloride hydrate is a hydrated form of iridium trichloride used as a research-grade biochemical reagent and as a source of iridium for synthesis, catalysis, and bioinorganic studies. The material is supplied as a crystalline hydrated chloride with recommended storage and handling conditions for laboratory use.

  • Hydrated formula: IrCl3 · xH2O.
  • CAS number: 14996-61-3.
  • Molecular weight: 298.58 g/mol (anhydrous basis).
  • Purity typically 98%.
  • Available in small pack sizes, including 1 g.
  • Store protected from light at 4°C; in solution store at -20°C to -80°C as recommended.
  • Used as a source of iridium for synthesis, catalysis, and biomedical research.

Catalog No. 50-003-00115 Supplier Medchemexpress LLC Supplier No. HYW0210221G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Iridium(III) chloride hydrate is a hydrated form of iridium trichloride used as a research-grade biochemical reagent and as a source of iridium for synthesis, catalysis, and bioinorganic studies. The material is supplied as a crystalline hydrated chloride with recommended storage and handling conditions for laboratory use.

  • Hydrated formula: IrCl3 · xH2O.
  • CAS number: 14996-61-3.
  • Molecular weight: 298.58 g/mol (anhydrous basis).
  • Purity typically 98%.
  • Available in small pack sizes, including 1 g.
  • Store protected from light at 4°C; in solution store at -20°C to -80°C as recommended.
  • Used as a source of iridium for synthesis, catalysis, and biomedical research.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.