Learn More
TARGETMOL CHEMICALS INC Iriflophenone 3-C-glucoside
SDP
Catalog No. 502880747
Encompass_Preferred
Also available in 1 mL, 1 mg, 10 mg, 25 mg and bulk. Please contact Fisher for quotes. Iriflophenone 3-C-glucoside (Iriflophenone 3-C-beta-D-glucopyranoside),has antioxidant activity,isolated from Cyclopia genistoides. Purity 98.87%
Catalog No. 50-288-0747 Supplier TARGETMOL CHEMICALS INC Supplier No. T116765MG
Add to Cart
Add to Cart
Also available in 1 mL, 1 mg, 10 mg, 25 mg and bulk. Please contact Fisher for quotes. Iriflophenone 3-C-glucoside (Iriflophenone 3-C-beta-D-glucopyranoside),has antioxidant activity,isolated from Cyclopia genistoides. Purity 98.87%