Learn More
Medchemexpress LLC Iriflophenone | 52591-10-3 | MFCD20260852 | 98.1% | 246.22 g/mol | C13H10O5 | 10 MG
SDP
Catalog No. 5000289843
Encompass_Preferred
Iriflophenone is a plant-derived phenolic compound (C13H10O5, MW 246.22) isolated from Aquilaria sinensis and provided for laboratory research; it has been reported to stimulate proliferation of MCF-7 and T-47D human breast cancer cell lines.
  • Plant-derived phenolic compound isolated from Aquilaria sinensis.
  • Reported to stimulate proliferation of MCF-7 and T-47D human breast cancer cells.
  • High purity suitable for in vitro research (reported purity ~98.1%).
  • Available as solid or as ready-to-use DMSO solution for cell-based assays.
  • Characterized chemical profile with CAS 52591-10-3 and formula C13H10O5.
Catalog No. 50-002-89843 Supplier Medchemexpress LLC Supplier No. HYN401010MG
Iriflophenone is a plant-derived phenolic compound (C13H10O5, MW 246.22) isolated from Aquilaria sinensis and provided for laboratory research; it has been reported to stimulate proliferation of MCF-7 and T-47D human breast cancer cell lines.
  • Plant-derived phenolic compound isolated from Aquilaria sinensis.
  • Reported to stimulate proliferation of MCF-7 and T-47D human breast cancer cells.
  • High purity suitable for in vitro research (reported purity ~98.1%).
  • Available as solid or as ready-to-use DMSO solution for cell-based assays.
  • Characterized chemical profile with CAS 52591-10-3 and formula C13H10O5.