Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Irigenin | 548-76-5 | MFCD00597047 | 99.8% | 360.31 g/mol | C18H16O8 | 10 MG
SDP

Catalog No. 5000289509
Encompass_Preferred

Irigenin is an isoflavone research compound used to inhibit Extra Domain A (EDA)-integrin interactions and to study integrin-mediated signaling and apoptosis in vitro. It is supplied as a solid powder and as DMSO solutions for assay preparation.

  • Inhibits EDA-integrin interactions and is used in integrin and apoptosis research.
  • High purity (99.84%) suitable for research applications.
  • Molecular weight 360.31 g/mol; chemical formula C18H16O8.
  • CAS number 548-76-5 for unambiguous identification.
  • Available as solid powder and 10 mM solutions in DMSO for assay use.
  • Available in small research pack sizes including 10 MG.

Catalog No. 50-002-89509 Supplier Medchemexpress LLC Supplier No. HYN258710MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Irigenin is an isoflavone research compound used to inhibit Extra Domain A (EDA)-integrin interactions and to study integrin-mediated signaling and apoptosis in vitro. It is supplied as a solid powder and as DMSO solutions for assay preparation.

  • Inhibits EDA-integrin interactions and is used in integrin and apoptosis research.
  • High purity (99.84%) suitable for research applications.
  • Molecular weight 360.31 g/mol; chemical formula C18H16O8.
  • CAS number 548-76-5 for unambiguous identification.
  • Available as solid powder and 10 mM solutions in DMSO for assay use.
  • Available in small research pack sizes including 10 MG.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.