Learn More
Medchemexpress LLC HY-16562 100mg Medchemexpress, Irinotecan CAS:97682-44-5 Purity:>98%
SDP
Catalog No. 502029203
Encompass_Preferred
Medchemexpress, HY-16562 100mg Irinotecan CAS:97682-44-5 Irinotecan ((+)-Irinotecan) is a topoisomerase I inhibitor, preventing religation of the DNA strand by binding to topoisomerase I-DNA complex. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9203 Supplier Medchemexpress LLC Supplier No. HY16562100MG
Medchemexpress, HY-16562 100mg Irinotecan CAS:97682-44-5 Irinotecan ((+)-Irinotecan) is a topoisomerase I inhibitor, preventing religation of the DNA strand by binding to topoisomerase I-DNA complex. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.