Learn More
Apexbio Technology LLC Irinotecan(Synonyms: CPT-11, Campto, Camptosar, irinotecan hydrochloride, Irinotecan HCl, irinotecan hydrochloride trihydrate), 10mM (in 1mL DMSO), CAS: 97682-44-5.
SDP
Supplier:  Apexbio Technology LLC A51335.1
Encompass_Preferred
Irinotecan (CAS 97682-44-5) also known as CPT-11 is an anticancer prodrug that acts as an inhibitor of topoisomerase I Upon enzymatic activation by carboxylesterase (CCE) to its potent metabolite SN-38 it stabilizes the DNA-topoisomerase I cleavable complex causing DNA damage and apoptosis In vitro studies show Irinotecan inhibits various colorectal cancer cell lines including LoVo (IC50 15 8 M) and HT-29 (IC50 5 17 M) Additionally xenograft models such as COLO 320 demonstrate pronounced tumor growth suppression Irinotecan is widely utilized in research to investigate DNA damage mechanisms and therapeutic efficacy in colorectal cancer
Catalog No. 50-000-60803