Learn More
Medchemexpress LLC Irinotecan-d10 | 718613-28-6 | 596.74 g/mol | C33H28D10N4O6 | 10 MG
SDP
Catalog No. 5000289309
Encompass_Preferred
Irinotecan-d10 is a deuterium-labeled isotopologue of irinotecan used as an analytical/internal standard in mass spectrometry and related assays. It serves as a research reference for studies involving topoisomerase I inhibition and quantitative bioanalysis.
  • Deuterium-labeled (d10) isotopologue for clear mass separation in MS.
  • Suitable as an internal standard for LC-MS/MS quantification.
  • Provides reliable mass shift for analyte tracking and recovery assessment.
  • Supplied as a small-molecule reference for research use only.
  • CAS number 718613-28-6 and molecular weight ~596.74 g/mol aid compound identification.
Catalog No. 50-002-89309 Supplier Medchemexpress LLC Supplier No. HY16562S10MG
Irinotecan-d10 is a deuterium-labeled isotopologue of irinotecan used as an analytical/internal standard in mass spectrometry and related assays. It serves as a research reference for studies involving topoisomerase I inhibition and quantitative bioanalysis.
  • Deuterium-labeled (d10) isotopologue for clear mass separation in MS.
  • Suitable as an internal standard for LC-MS/MS quantification.
  • Provides reliable mass shift for analyte tracking and recovery assessment.
  • Supplied as a small-molecule reference for research use only.
  • CAS number 718613-28-6 and molecular weight ~596.74 g/mol aid compound identification.