Learn More
Medchemexpress LLC Irinotecan-d10 | 718613-28-6 | 98.0% | 596.74 | 1 MG
SDP
Catalog No. 5000391764
Encompass_Preferred
Irinotecan-d10 is a deuterium-labeled Irinotecan, which functions as a topoisomerase I inhibitor. It prevents the religation of DNA strands by binding to the topoisomerase I-DNA complex, making it suitable for various research applications.
  • Deuterium-labeled compound for research use
  • Acts as a topoisomerase I inhibitor
  • Prevents religation of DNA strands
  • Functions as a tracer
  • Suitable as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Store at 4°C, protected from light
  • For longer storage in solvent, keep at -80°C for 6 months or -20°C for 1 month, protected from light
Catalog No. 50-003-91764 Supplier Medchemexpress LLC Supplier No. HY16562S1MG
Irinotecan-d10 is a deuterium-labeled Irinotecan, which functions as a topoisomerase I inhibitor. It prevents the religation of DNA strands by binding to the topoisomerase I-DNA complex, making it suitable for various research applications.
  • Deuterium-labeled compound for research use
  • Acts as a topoisomerase I inhibitor
  • Prevents religation of DNA strands
  • Functions as a tracer
  • Suitable as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Store at 4°C, protected from light
  • For longer storage in solvent, keep at -80°C for 6 months or -20°C for 1 month, protected from light