Learn More
Medchemexpress LLC HY-16562A 500mg , Irinotecan (hydrochloride) CAS:100286-90-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY16562A500MG
Encompass_Preferred
HY-16562A 500mg Irinotecan (hydrochloride) CAS: 100286-90-6 Irinotecan hydrochloride ((+)-Irinotecan hydrochloride) is a topoisomerase I inhibitor mainly used to treat colon cancer and rectal cancer. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9767