Learn More
Selleck Chemical LLC Irinotecan HCl Trihydrate S2217-100mg
SDP
Catalog No. 503112787
Encompass_Preferred
Irinotecan HCl Trihydrate is a hydrochloride trihydrate of irinotecan (Camptosar Campto CPT-11) which is a topoisomerase I inhibitor with IC50 of 15 8 and 5 17 M for LoVo cells and HT-29 cells respectively
Catalog No. 50-311-2787 Supplier Selleck Chemical LLC Supplier No. S2217100MG
Add to Cart
Add to Cart
Irinotecan HCl Trihydrate is a hydrochloride trihydrate of irinotecan (Camptosar Campto CPT-11) which is a topoisomerase I inhibitor with IC50 of 15 8 and 5 17 M for LoVo cells and HT-29 cells respectively