Learn More
Medchemexpress LLC Irinotecan hydrochloride | 100286-90-6 | 99.9% | 1 ML
SDP
Catalog No. 5000393895
Encompass_Preferred
Irinotecan hydrochloride is a topoisomerase I inhibitor primarily used to treat colon cancer and rectal cancer. It is intended for research use only.
  • Acts as a topoisomerase I inhibitor
  • Inhibits tumor growth in various models
  • Induces cell cycle arrest and apoptosis
  • Suitable for in vitro and in vivo studies
Catalog No. 50-003-93895 Supplier Medchemexpress LLC Supplier No. HY16562A10MM/1ML
Irinotecan hydrochloride is a topoisomerase I inhibitor primarily used to treat colon cancer and rectal cancer. It is intended for research use only.
  • Acts as a topoisomerase I inhibitor
  • Inhibits tumor growth in various models
  • Induces cell cycle arrest and apoptosis
  • Suitable for in vitro and in vivo studies