Learn More
Selleck Chemical LLC Irinotecan hydrochloride S5026-10mM/1mL
SDP
Catalog No. 503112452
Encompass_Preferred
Irinotecan (CPT-11 ( )-Irinotecan) hydrochloride is an inhibitor of Topoisomerase I (Topo I) that exhibits cytotoxicity in LoVo cells and HT-29 cells with IC50 of 15 8 M and 5 17 M respectively
Catalog No. 50-311-2452 Supplier Selleck Chemical LLC Supplier No. S502610MM/1ML
Irinotecan (CPT-11 ( )-Irinotecan) hydrochloride is an inhibitor of Topoisomerase I (Topo I) that exhibits cytotoxicity in LoVo cells and HT-29 cells with IC50 of 15 8 M and 5 17 M respectively