Learn More
Medchemexpress LLC HY-16568 50mg Medchemexpress, Irinotecan (hydrochloride trihydrate) CAS:136572-09-3 Purity:>98%
SDP
Catalog No. 502030725
Encompass_Preferred
Medchemexpress, HY-16568 50mg Irinotecan (hydrochloride trihydrate) CAS:136572-09-3 Irinotecan hydrochloride trihydrate ((+)-Irinotecan hydrochloride trihydrate) is a topoisomerase I inhibitor with antitumor activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0725 Supplier Medchemexpress LLC Supplier No. HY1656850MG
Medchemexpress, HY-16568 50mg Irinotecan (hydrochloride trihydrate) CAS:136572-09-3 Irinotecan hydrochloride trihydrate ((+)-Irinotecan hydrochloride trihydrate) is a topoisomerase I inhibitor with antitumor activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.