Learn More
Selleck Chemical LLC Irinotecan hydrochloride 5mg
SDP
Supplier:  Selleck Chemical LLC S50265MG
Encompass_Preferred
Irinotecan (CPT-11, (+)-Irinotecan) hydrochloride is an inhibitor of Topoisomerase I (Topo I) that exhibits cytotoxicity in LoVo cells and HT-29 cells with IC50 of 15.8 ?M and 5.17 ?M, respectively. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-217-3153