Learn More
Medchemexpress LLC Irinotecan hydrochloride trihydrate | 136572-09-3 | 99.9% | 200 MG
SDP
Catalog No. 5000393835
Encompass_Preferred
Irinotecan hydrochloride trihydrate is a potent topoisomerase I inhibitor known for its antitumor activity. It targets Topoisomerase I, playing a crucial role in cancer research and drug development.
  • Inhibits the growth of LoVo and HT-29 cells.
  • Suppresses the proliferation of human umbilical vein endothelial cells (HUVEC).
  • Significantly inhibits tumor growth in various in vivo models.
  • Shows effectiveness against HT-29 xenografts.
Catalog No. 50-003-93835 Supplier Medchemexpress LLC Supplier No. HY16568200MG
Irinotecan hydrochloride trihydrate is a potent topoisomerase I inhibitor known for its antitumor activity. It targets Topoisomerase I, playing a crucial role in cancer research and drug development.
  • Inhibits the growth of LoVo and HT-29 cells.
  • Suppresses the proliferation of human umbilical vein endothelial cells (HUVEC).
  • Significantly inhibits tumor growth in various in vivo models.
  • Shows effectiveness against HT-29 xenografts.