Learn More
Medchemexpress LLC Irinotecan hydrochloride trihydrate | 136572-09-3 | 99.9% | 677.18 | 1 ML
SDP
Catalog No. 5000394187
Encompass_Preferred
Irinotecan hydrochloride, also known as (+)-Irinotecan hydrochloride or CPT-11 hydrochloride, is a topoisomerase I inhibitor primarily used in the treatment of colon and rectal cancer. This product is intended for research use only.
  • Acts as an inhibitor of topoisomerase I, an enzyme crucial for DNA replication, transcription, and repair.
  • Effectively inhibits the growth of various cancer cell lines.
  • Suppresses the proliferation of human umbilical vein endothelial cells (HUVEC).
  • Significantly inhibits tumor growth in animal models.
  • Involved in numerous clinical trials for a wide range of cancers.
Catalog No. 50-003-94187 Supplier Medchemexpress LLC Supplier No. HY1656810MM/1ML
Irinotecan hydrochloride, also known as (+)-Irinotecan hydrochloride or CPT-11 hydrochloride, is a topoisomerase I inhibitor primarily used in the treatment of colon and rectal cancer. This product is intended for research use only.
  • Acts as an inhibitor of topoisomerase I, an enzyme crucial for DNA replication, transcription, and repair.
  • Effectively inhibits the growth of various cancer cell lines.
  • Suppresses the proliferation of human umbilical vein endothelial cells (HUVEC).
  • Significantly inhibits tumor growth in animal models.
  • Involved in numerous clinical trials for a wide range of cancers.