Learn More
Medchemexpress LLC Irinotecan hydrochloride trihydrate | 136572-09-3 | 99.9% | 677.18 | 100 MG
SDP
Catalog No. 5000393871
Encompass_Preferred
Irinotecan hydrochloride trihydrate is a topoisomerase I inhibitor with antitumor activity, used to treat colon and rectal cancer. It is for research use only and has been shown to inhibit the growth of various cancer cell lines, such as LoVo and HT-29.
  • Inhibits topoisomerase I
  • Demonstrates antitumor activity
  • Suppresses proliferation of human umbilical vein endothelial cells
  • Available for research use only
Catalog No. 50-003-93871 Supplier Medchemexpress LLC Supplier No. HY16568100MG
Irinotecan hydrochloride trihydrate is a topoisomerase I inhibitor with antitumor activity, used to treat colon and rectal cancer. It is for research use only and has been shown to inhibit the growth of various cancer cell lines, such as LoVo and HT-29.
  • Inhibits topoisomerase I
  • Demonstrates antitumor activity
  • Suppresses proliferation of human umbilical vein endothelial cells
  • Available for research use only