Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Irinotecan Hydrochloride Trihydrate 98.0+%, TCI America™
SDP

Catalog No. I0714100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I0714100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I0714100MG Supplier TCI America Supplier No. I0714100MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 136572-09-3
Molecular Formula C33H45ClN4O9
Molecular Weight (g/mol) 677.192
MDL Number MFCD01765731
InChI Key KLEAIHJJLUAXIQ-JDRGBKBRSA-N
Synonym irinotecan hydrochloride trihydrate, irinotecan hcl trihydrate, cpt 11, unii-042laq1iis, campto, irinotecan hcl, 042laq1iis, onivyde, cpt-11
PubChem CID 60837
SMILES CCC1=C2C=C(C=CC2=NC3=C1CN4C3=CC5=C(C4=O)COC(=O)C5(CC)O)OC(=O)N6CCC(CC6)N7CCCCC7.O.O.O.Cl

Specifications

Boiling Point 257°C
Quantity 100 mg
Formula Weight 623.15
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Irinotecan Hydrochloride Trihydrate
RTECSNumber DW1060750
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.