Learn More
Cayman Chemical Irinotecan (hydrochloride hydrate), Purity: ≥98%, CAS: 136572-09-3, 100 mg
SDP
Catalog No. 502955523
Encompass_Preferred
A derivative of the alkaloid camptothecin that is converted by tissue carboxylesterase to 7-ethyl-10-hydroxycamptothecin, a potent inhibitor of DNA topoisomerase I; demonstrates a broad spectrum of antitumor activity against metastatic colorectal cancer, small cell lung cancer, and several other solid tumors
Catalog No. 50-295-5523 Supplier Cayman Chemical Supplier No. 14180100 MG
A derivative of the alkaloid camptothecin that is converted by tissue carboxylesterase to 7-ethyl-10-hydroxycamptothecin, a potent inhibitor of DNA topoisomerase I; demonstrates a broad spectrum of antitumor activity against metastatic colorectal cancer, small cell lung cancer, and several other solid tumors