Learn More
Electron Microscopy Sciences Iris Dressing Forceps Iris Dressing Forceps Full Curved
SDP
Catalog No. 5030209
Encompass_Preferred
Made from stainless steel, 4" long.The forceps are delicate and the tips are serrated (1 mm wide). Comes complete with a guide pin.
Catalog No. 50-302-09 Supplier Electron Microscopy Sciences Supplier No. 786104FC
Made from stainless steel, 4" long.The forceps are delicate and the tips are serrated (1 mm wide). Comes complete with a guide pin.