Learn More
Electron Microscopy Sciences IRIS – Dressing Forceps, Straight
SDP
Catalog No. 5030208
Encompass_Preferred
Made from stainless steel, 4" long.The forceps are delicate and the tips are serrated (1 mm wide). Comes complete with a guide pin.
Catalog No. 50-302-08 Supplier Electron Microscopy Sciences Supplier No. 786104S
Made from stainless steel, 4" long.The forceps are delicate and the tips are serrated (1 mm wide). Comes complete with a guide pin.