Learn More
World Precision Instrument Iris Forceps, 10cm, Straight, Serrated, 12-pack
SDP
Catalog No. 50379971
Encompass
Iris Forceps are a pair of surgical forceps (tweezers) often used in ophthalmic procedures. Iris Forceps are a small pair of forceps with a fine pointed tip for grasping delicate tissue with minimal trauma. The narrow, serrated jaws of the surgical forceps allow for fine manipulation of structures like the iris of the eye. Researchers like these fine tipped tweezers for precision work in microdissection. These forceps are 10cm long with fine tips, and they come with straight or curved tips. The Iris Forceps are manufactures with surgical grade stainless steel. Our Disposable Iris Forceps are designed to withstand the high heat and chemical exposure of the sterilization process. They may be autoclaved. These economy forceps are sold in packages of 12, perfect for students or for disposable use. All sales are final on disposable instrument packs.
Catalog No. 50-379-971 Supplier World Precision Instrument Supplier No. 50372112
 Restricted Item
Add to Cart
Add to Cart
Iris Forceps are a pair of surgical forceps (tweezers) often used in ophthalmic procedures. Iris Forceps are a small pair of forceps with a fine pointed tip for grasping delicate tissue with minimal trauma. The narrow, serrated jaws of the surgical forceps allow for fine manipulation of structures like the iris of the eye. Researchers like these fine tipped tweezers for precision work in microdissection. These forceps are 10cm long with fine tips, and they come with straight or curved tips. The Iris Forceps are manufactures with surgical grade stainless steel. Our Disposable Iris Forceps are designed to withstand the high heat and chemical exposure of the sterilization process. They may be autoclaved. These economy forceps are sold in packages of 12, perfect for students or for disposable use. All sales are final on disposable instrument packs.