Learn More
Fine Science Tools IRIS FORCEPS, STR, 1X2 TEETH
SDP
Catalog No. NC0545063
Encompass
Tip Shape: Straight/ Tips: 1x2 Teeth/ Tip Dimensions: 0.6 x 0.5mm/ Alloy / Material: Stainless Steel/ Length: 7cm
Catalog No. NC0545063 Supplier Fine Science Tools Supplier No. 1106607
Tip Shape: Straight/ Tips: 1x2 Teeth/ Tip Dimensions: 0.6 x 0.5mm/ Alloy / Material: Stainless Steel/ Length: 7cm