Learn More
Med Vet International Miltex Vantage Iris Forceps, 4in, Full Curved, with 1x2 Teeth
Supplier:  Med Vet International MILV918790
Encompass_Preferred
The Miltex Vantage V918-790 Iris forceps are made from high quality Japanese stainless steel and measure 4' in length. They have a non-locking thumb handle and full-curved tips with 1 x 2 teeth.
Catalog No. 50-255-9468