Learn More
Med Vet International Miltex Vantage Iris Scissors, 4.125in., Curved
Catalog No. 502830575
Encompass_Preferred
The Miltex Vantage V95-306 iris scissors are made from high quality Japanese stainless steel and measure 4.125" in length. They have a finger ring handle and curved blades with sharp tips.
Catalog No. 50-283-0575 Supplier Med Vet International Supplier No. MILV95306
The Miltex Vantage V95-306 iris scissors are made from high quality Japanese stainless steel and measure 4.125" in length. They have a finger ring handle and curved blades with sharp tips.