Learn More
Fine Science Tools Fine Scissors, Sharp, Stainless Steel, Curved, 11.5 cm
SDP
Catalog No. NC9518521
Encompass
Tip Shape: Curved/ Tips: Sharp-Sharp/ Cutting Edge: 25mm/ Alloy / Material: Stainless Steel/ Length: 11.5cm/ Serrated: No
Catalog No. NC9518521 Supplier Fine Science Tools Supplier No. 1406111
Tip Shape: Curved/ Tips: Sharp-Sharp/ Cutting Edge: 25mm/ Alloy / Material: Stainless Steel/ Length: 11.5cm/ Serrated: No