Learn More
World Precision Instrument Iris Scissors, 11.5cm, SuperCut, Straight
SDP
Catalog No. NC9576989
Encompass
Iris scissors were originally designed for ophthalmic procedures but are now used in a wide range of applications. The tips are sharp, and they come in curved, straight or angled styles.
Catalog No. NC9576989 Supplier World Precision Instrument Supplier No. 503259
Iris scissors were originally designed for ophthalmic procedures but are now used in a wide range of applications. The tips are sharp, and they come in curved, straight or angled styles.