Learn More
Fine Science Tools Bonn Artery Scissors, Ball Tip, Stainless Steel, 9 cm
SDP
Catalog No. NC9173725
Encompass
Tip Shape: Straight/ Tips: Sharp-Ball Tip/ Cutting Edge: 13mm/ Alloy / Material: Stainless Steel/ Length: 9cm/ Serrated: No/ Feature: 1.5mm Ball Diameter
Catalog No. NC9173725 Supplier Fine Science Tools Supplier No. 1408609
Tip Shape: Straight/ Tips: Sharp-Ball Tip/ Cutting Edge: 13mm/ Alloy / Material: Stainless Steel/ Length: 9cm/ Serrated: No/ Feature: 1.5mm Ball Diameter