Learn More
World Precision Instrument Iris Scissors, 10.5cm, Curved, Sharp, Titanium
SDP
Catalog No. 502538306
Encompass
Iris scissors were originally designed for ophthalmic procedures but are now used in a wide range of applications. The tips are sharp, and they come in curved, straight or angled styles.
Catalog No. 50-253-8306 Supplier World Precision Instrument Supplier No. WP4410L
Iris scissors were originally designed for ophthalmic procedures but are now used in a wide range of applications. The tips are sharp, and they come in curved, straight or angled styles.