Learn More
World Precision Instrument Iris Scissors, 12.5cm, SuperCut, Straight, German
SDP
Catalog No. 502539557
Encompass
Iris scissors were originally designed for ophthalmic procedures but are now used in a wide range of applications. The tips are sharp, and they come in curved, straight or angled styles.
Catalog No. 50-253-9557 Supplier World Precision Instrument Supplier No. 14225G
Iris scissors were originally designed for ophthalmic procedures but are now used in a wide range of applications. The tips are sharp, and they come in curved, straight or angled styles.