Learn More
Sklar Instruments Iris Scissors, 3-1/2 in., Sharp/Sharp, Straight, Smooth, Stainless Steel, EA.
SDP
Catalog No. 501109761 Shop All Sklar Instruments Products
Encompass_Preferred
Iris Scissors, 3-1/2 in., Sharp/Sharp, Straight, Smooth, Stainless Steel, EA. Originally designed for ophthalmic procedures, Iris Scissors have become a multipurpose instrument used in various surgical environments. These fine, sharp scissors are ideal for detailed dissection of fine tissue and fine suture removal. In addition to ophthalmic procedures, Iris Scissors are now often used in emergency rooms and for OB/GYN and dermatologic procedures. This product is straight with smooth, sharp/sharp tips and a length of 3-1/2 inches.
Catalog No. 50-110-9761 Supplier Sklar Instruments Supplier No. 471135
Iris Scissors, 3-1/2 in., Sharp/Sharp, Straight, Smooth, Stainless Steel, EA. Originally designed for ophthalmic procedures, Iris Scissors have become a multipurpose instrument used in various surgical environments. These fine, sharp scissors are ideal for detailed dissection of fine tissue and fine suture removal. In addition to ophthalmic procedures, Iris Scissors are now often used in emergency rooms and for OB/GYN and dermatologic procedures. This product is straight with smooth, sharp/sharp tips and a length of 3-1/2 inches.