Learn More
Sklar Instruments Econo Iris Scissors, Sklar, 3-1/2 in., Sharp/Sharp, Curved, Smooth, Finger Rings, Stainless Steel. Box of 12.
SDP
Catalog No. 501437537 Shop All Sklar Instruments Products
Encompass_Preferred
Econo Iris Scissors, Sklar, 3-1/2 in., Sharp/Sharp, Curved, Smooth, Finger Rings, Stainless Steel. Originally designed for ophthalmic procedures, Iris Scissors have become a multipurpose instrument used in various surgical environments. These fine, sharp scissors are ideal for detailed dissection of fine tissue and fine suture removal. In addition to ophthalmic procedures, Iris Scissors are now often used in emergency rooms and for OB/GYN and dermatologic procedures. Econo instruments are Sklar's floor-grade, disposable instruments and are sold in bulk quantities. This product is curved with smooth edges, sharp tips, and a length of 3-1/2 inches. They are available in a box of 12.
This item is not returnable. View return policy
Catalog No. 50-143-7537 Supplier Sklar Instruments Supplier No. 21107
Econo Iris Scissors, Sklar, 3-1/2 in., Sharp/Sharp, Curved, Smooth, Finger Rings, Stainless Steel. Originally designed for ophthalmic procedures, Iris Scissors have become a multipurpose instrument used in various surgical environments. These fine, sharp scissors are ideal for detailed dissection of fine tissue and fine suture removal. In addition to ophthalmic procedures, Iris Scissors are now often used in emergency rooms and for OB/GYN and dermatologic procedures. Econo instruments are Sklar's floor-grade, disposable instruments and are sold in bulk quantities. This product is curved with smooth edges, sharp tips, and a length of 3-1/2 inches. They are available in a box of 12.