Learn More
Med Vet International Miltex Vantage Iris Scissors, 4.125in, Straight
Catalog No. 502522272
Encompass_Preferred
The Miltex Vantage V95-304 iris scissors are made from high quality Japanese stainless steel and measure 4.125" in length. They have a finger ring handle and straight blades with sharp tips.
Catalog No. 50-252-2272 Supplier Med Vet International Supplier No. MILV95304
The Miltex Vantage V95-304 iris scissors are made from high quality Japanese stainless steel and measure 4.125" in length. They have a finger ring handle and straight blades with sharp tips.