Learn More
Sklar Instruments Merit Sterile™ Iris Scissors - 4-1/2", Curved, Sharp/Sharp, Disposable, CS 25
SDP
Catalog No. NC3351376 Shop All Sklar Instruments Products
Encompass_Preferred
Merit Sterile™ Iris Scissors were originally designed for ophthalmic procedures, but have become a multipurpose instrument used in various surgical environments. These fine, sharp scissors are ideal for detailed dissection of fine tissue and fine suture removal. In addition to ophthalmic procedures, Iris Scissors are now often used multiple procedures. Merit Sterile™ instruments are ready-to-use, sterile, disposable, office-grade instruments sold in bulk quantities.
This item is not returnable. View return policy
Catalog No. NC3351376 Supplier Sklar Instruments Supplier No. 962507M
Merit Sterile™ Iris Scissors were originally designed for ophthalmic procedures, but have become a multipurpose instrument used in various surgical environments. These fine, sharp scissors are ideal for detailed dissection of fine tissue and fine suture removal. In addition to ophthalmic procedures, Iris Scissors are now often used multiple procedures. Merit Sterile™ instruments are ready-to-use, sterile, disposable, office-grade instruments sold in bulk quantities.