Learn More
Fine Science Tools Fine Scissors, Stainless Steel, Sharp, 10.5 cm
SDP
Catalog No. NC9550426
Encompass
Tip Shape: Curved to Side/ Tips: Sharp-Sharp/ Cutting Edge: 21mm/ Alloy / Material: Stainless Steel/ Length: 10.5cm/ Serrated: No
Catalog No. NC9550426 Supplier Fine Science Tools Supplier No. 1406210
Tip Shape: Curved to Side/ Tips: Sharp-Sharp/ Cutting Edge: 21mm/ Alloy / Material: Stainless Steel/ Length: 10.5cm/ Serrated: No