Learn More
Electron Microscopy Sciences EMS Tungsten Carbide Iris Scissors with Curved Blades, 4-1/2"
SDP
Catalog No. 501931430
Encompass_Preferred
These Iris scissors are made from Tungsten Carbide and have sharp/sharp points. The unique tungsten carbide cutting edge provides effortless, yet accurate, cuts through the toughest of surgical or autopsy tissue. Scissor shanks are tempered to the ideal hardness to allow proper flexibility and comfort. Finger rings are well rounded and polished allowing better feel while preventing finger fatigue during complex dissection cases.
Blade: Curved
OAL: 4-1/2"
Catalog No. 50-193-1430 Supplier Electron Microscopy Sciences Supplier No. 7299622
These Iris scissors are made from Tungsten Carbide and have sharp/sharp points. The unique tungsten carbide cutting edge provides effortless, yet accurate, cuts through the toughest of surgical or autopsy tissue. Scissor shanks are tempered to the ideal hardness to allow proper flexibility and comfort. Finger rings are well rounded and polished allowing better feel while preventing finger fatigue during complex dissection cases.
Blade: Curved
OAL: 4-1/2"