Learn More
Fine Science Tools Fine Scissors - Sharp, 10.5cm
SDP
Catalog No. NC9691799
Encompass
Tip Shape: Curved/ Tips: Sharp-Sharp/ Cutting Edge: 21mm/ Alloy / Material: Stainless Steel/ Length: 10.5cm/ Serrated: No
Catalog No. NC9691799 Supplier Fine Science Tools Supplier No. 1406110
Tip Shape: Curved/ Tips: Sharp-Sharp/ Cutting Edge: 21mm/ Alloy / Material: Stainless Steel/ Length: 10.5cm/ Serrated: No