Learn More
Fine Science Tools Noyes Spring Scissors, Stainless Steel, Straight, 12 cm
SDP
Catalog No. NC9019530
Encompass
Tip Shape: Straight/ Tips: Sharp/ Cutting Edge: 14mm/ Tip Diameter: 0.275mm/ Alloy / Material: Stainless Steel/ Length: 12cm/ Serrated: No
Catalog No. NC9019530 Supplier Fine Science Tools Supplier No. 1501212
Tip Shape: Straight/ Tips: Sharp/ Cutting Edge: 14mm/ Tip Diameter: 0.275mm/ Alloy / Material: Stainless Steel/ Length: 12cm/ Serrated: No