Learn More
Braintree Scientific Iris Scissors, Standard Pattern,Straight, 4.5""""
SDP
Catalog No. NC0447593
Encompass
Iris Scissors, Standard Pattern, for precise vascular and neurological dissection. Ideal for small specimens. 4 1/2" (114.3mm) **Sold for research purposes only and not for human use**
Catalog No. NC0447593 Supplier Braintree Scientific Supplier No. SCI128
Iris Scissors, Standard Pattern, for precise vascular and neurological dissection. Ideal for small specimens. 4 1/2" (114.3mm) **Sold for research purposes only and not for human use**