Learn More
Electron Microscopy Sciences EMS SuperCut Iris Scissors with Straight Blades, 4-1/2"
SDP
Catalog No. 501931416
Encompass_Preferred
These Iris blade scissors have a specially designed sharp end. Blades have a curve to attain the hard to reach delicate tissue or angle dissections. SuperCut scissors are made in Germany using the highest grade of surgical steel and are tempered and hardened for an extended and sharper life than ordinary steel. Rounded and polished finger rings
Blade: Straight
OAL: 4-1/2"
This item is not returnable. View return policy
Catalog No. 50-193-1416 Supplier Electron Microscopy Sciences Supplier No. 7299609
These Iris blade scissors have a specially designed sharp end. Blades have a curve to attain the hard to reach delicate tissue or angle dissections. SuperCut scissors are made in Germany using the highest grade of surgical steel and are tempered and hardened for an extended and sharper life than ordinary steel. Rounded and polished finger rings
Blade: Straight
OAL: 4-1/2"