Learn More
Electron Microscopy Sciences EMS Iris Scissors Standard with Anglular P{robe Points
SDP
Catalog No. 501931299
Encompass_Preferred
These Iris scissors are made with sharp tips and are perfect for precise vascular and neurological dissection; ideal for small specimens and animal dissection.
OAL: 114 mm
Catalog No. 50-193-1299 Supplier Electron Microscopy Sciences Supplier No. 7299815
These Iris scissors are made with sharp tips and are perfect for precise vascular and neurological dissection; ideal for small specimens and animal dissection.
OAL: 114 mm