Learn More
Fine Science Tools Iris Spatulae, Tip: 20 x 1.8 x 0.3 mm, Stainless Steel, 12 cm
SDP
Catalog No. NC9493062
Encompass
Individual Item Name: Iris Spatulae - Curved/12cm/ Tip Shape: Curved/ Tip Dimensions: 20 x 1.8 x 0.3mm/ Alloy: Stainless Steel/ Length: 12cm
Catalog No. NC9493062 Supplier Fine Science Tools Supplier No. 1009212
Individual Item Name: Iris Spatulae - Curved/12cm/ Tip Shape: Curved/ Tip Dimensions: 20 x 1.8 x 0.3mm/ Alloy: Stainless Steel/ Length: 12cm