Learn More
Fine Science Tools Iris Spatulae
SDP
Catalog No. NC1913874
Encompass
Iris Spatulae, Alloy / Material: Stainless Steel, Length: 13cm, Tip Dimensions: 23 x 1.7 x 0.3mm, Tip Shape: Slight Curve
Catalog No. NC1913874 Supplier Fine Science Tools Supplier No. 1009313
Iris Spatulae, Alloy / Material: Stainless Steel, Length: 13cm, Tip Dimensions: 23 x 1.7 x 0.3mm, Tip Shape: Slight Curve