Learn More
Electron Microscopy Sciences Iris Tissue Forceps, Half Curved
SDP
Catalog No. 5030215
Encompass_Preferred
With 1x2 teeth they are extra delicate. Made from stainless steel and 4" long. The tips are 1.2 mm wide.
Catalog No. 50-302-15 Supplier Electron Microscopy Sciences Supplier No. 786114HC
With 1x2 teeth they are extra delicate. Made from stainless steel and 4" long. The tips are 1.2 mm wide.