Learn More
Electron Microscopy Sciences Iris 1x2 Teeth Tissue Forceps, Straight
SDP
Catalog No. 5030213
Encompass_Preferred
With 1x2 teeth they are extra delicate. Made from stainless steel and 4" long. The tips are 1.2 mm wide.
Catalog No. 50-302-13 Supplier Electron Microscopy Sciences Supplier No. 786114S
With 1x2 teeth they are extra delicate. Made from stainless steel and 4" long. The tips are 1.2 mm wide.